Learn More
Hamilton™ Micro Pipet Disposable Tips
Catalog No. 13684112B Shop All Hamilton Company Products
Change view
Click to view available options
For Use With (Equipment):
Microsyringe Pipet Model 701
Catalog No. For Use With (Equipment)
13-684-112B Microsyringe Pipet Model 701
1 options
Catalog No. 13-684-112B Supplier Hamilton™ Supplier No. 84255
Add to Cart
Add to Cart
Microsyringe Pipets have special 3/4 in. electrotapered needle for easy attachment of PTFE tips
  • Disposable PTFE tips that are to be used with the Microsyringe Pipet (13-684-112A)
  • Sample only wets disposable PTFE tips, eliminating sample cross-contamination
  • Spring-loaded plunger and adjustable stops ensure excellent reproducibility
  • Adjustable stop allows transfer of samples from .2 to 3μL
Specifications
Material PTFE
Disposable Single-use
Volume (Metric) 0.2 to 3 μL
For Use With (Equipment) Microsyringe Pipet Model 701