Learn More
Hamilton™ Microlab 300 Pipetting System Accessories - ClickSure Tips
Catalog No. p-4511774 Shop All Hamilton Company Products
Change view
Click to view available options
Volume (Metric):
50 μL
1 mL
Sterility:
Non-sterile
Sterile
Packaging Type:
Bulk
Rack
Racked
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Packaging Type
14-815-931 50 μL Sterile Bulk
14-815-930 50 μL Sterile Racked
14-815-935 1 mL Sterile Racked
14-815-934 1 mL Sterile Rack
14-815-933 1 mL Non-sterile Racked
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14-815-931 Supplier Hamilton™ Supplier No. 235544
Unique, environmentally friendly tips offer superior user performance
For use with Hamilton Microlab 300 Pipettor hand probe. When tips are placed onto the probe, the O-ring inside the probe compresses. This low insertion force results in a more ergonomic experience for the user that enables long-term use with minimal energy exerted.
Features and benefits:
  • Two tips, 50μL and 1mL, cover the entire pipetting volume range which means less inventory to store
  • Tip packaging uses 37% less plastic than leading competitor's packaging, saving on shipping weight and fossil fuels
  • Tip packaging and tips are recyclable for eco-conscious labs
  • Packaging is RoHS-compliant
  • Utility-grade recycled plastic is used for the tip packaging
  • Tips are chlorofluorocarbons (CFC)-free and cause no impact to the ozone
  • Manufacturing process consumes less energy and leads to more savings for you
Specifications
Sterility Sterile
Volume (Metric) 50 μL
For Use With (Equipment) Microlab™ 300 pipette
Packaging Type Bulk