Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Accessory, AirShield™ Protection Device
Catalog No. 13622251 Shop All Hamilton Company Products
Change view
Click to view available options
For Use With (Equipment):
For Microlab 600 Diluter
Catalog No. For Use With (Equipment)
13-622-251 For Microlab 600 Diluter
1 options
Catalog No. 13-622-251 Supplier Hamilton™ Supplier No. 6856201
Add to Cart
Add to Cart
Provides protection from harsh environmental conditions when used with Microlab 600™ Diluter/Dispenser System
  • Creates a positive pressure blanket of fresh air inside the pump that pushes air out over critical components for extra protection
  • Provides 24/7 protection with an independent power source
  • Extends the time between and reduces the cost of product maintenance
  • Compatible with all Microlab 600 series instruments
Specifications
Product Type AirShield
For Use With (Equipment) For Microlab 600 Diluter