Learn More
Analytical Sales & Services Hamilton Model 1001 1mL Removable Needle Gastight Syringe
Catalog No. 5000804318
Encompass
Hamilton Model 1001 1mL Removable Needle Gastight Syringe - 22 Gauge, 2" Length, Point Style #3
Catalog No. 50-008-04318 Supplier Analytical Sales & Services Supplier No. 4563184
Hamilton Model 1001 1mL Removable Needle Gastight Syringe - 22 Gauge, 2" Length, Point Style #3