Learn More
Analytical Sales & Services Hamilton Model 1725 N 250µL CTC Syringe w Cemented Needle
Catalog No. 5000804481
Encompass
Hamilton Model 1725 N 250µL CTC Syringe w Cemented Needle, 22 Gauge, 51mm Length, Point Style #3
Qty/Pkg: EACH
Catalog No. 50-008-04481 Supplier Analytical Sales & Services Supplier No. 42030794
Hamilton Model 1725 N 250µL CTC Syringe w Cemented Needle, 22 Gauge, 51mm Length, Point Style #3
Qty/Pkg: EACH