Learn More
Analytical Sales & Services Hamilton Model 1750 LTN 500 µL Gastight Syringe Cemented Needle
Catalog No. 5000807212
Encompass
Hamilton Model 1750 LTN 500 µL Gastight Syringe w Cemented Needle, 22 Gauge, 2" Length, Point Style #3
Qty/Pkg: EACH
Catalog No. 50-008-07212 Supplier Analytical Sales & Services Supplier No. 4612184
Hamilton Model 1750 LTN 500 µL Gastight Syringe w Cemented Needle, 22 Gauge, 2" Length, Point Style #3
Qty/Pkg: EACH