Learn More
Analytical Sales & Services Hamilton Model 701N 10µL Cemented Needle Microliter Syringe
Catalog No. 5000807038
Encompass
Hamilton Model 701N 10µL Cemented Needle Microliter Syringe, 26s Gauge, 2" Length, Point Style #2
Qty/Pkg: EACH
Catalog No. 50-008-07038 Supplier Analytical Sales & Services Supplier No. 4803004
Hamilton Model 701N 10µL Cemented Needle Microliter Syringe, 26s Gauge, 2" Length, Point Style #2
Qty/Pkg: EACH