Learn More
Analytical Sales & Services Hamilton Model 725N 250µL Cemented Needle Microliter Syringe
Catalog No. 5000806919
Encompass
Hamilton Model 725N 250µL Cemented Needle Microliter Syringe, 22s Gauge, 2" Length, Point Style #2
Qty/Pkg: EACH
Catalog No. 50-008-06919 Supplier Analytical Sales & Services Supplier No. 4807004
Hamilton Model 725N 250µL Cemented Needle Microliter Syringe, 22s Gauge, 2" Length, Point Style #2
Qty/Pkg: EACH