Learn More
Analytical Sales & Services Hamilton Model 750N 500µL Cemented Needle Microliter Syringe
Catalog No. 5000804910
Encompass
Hamilton Model 750N 500µL Cemented Needle Microliter Syringe, 22s Gauge, 2" Length, Point Style #2
Qty/Pkg: EACH
Catalog No. 50-008-04910 Supplier Analytical Sales & Services Supplier No. 4808004
Hamilton Model 750N 500µL Cemented Needle Microliter Syringe, 22s Gauge, 2" Length, Point Style #2
Qty/Pkg: EACH