Learn More
Hamilton™ Preflanged PTFE Tubing Assemblies
Catalog No. 14819167 Shop All Hamilton Company Products
Change view
Click to view available options
Product Type:
Tubing Assembly; 1/4-28 NT (SR)
Tubing Assembly; 1/4-28 TP (SR)
Overall Length (Metric):
762 mm
Gauge:
18 ga
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Overall Length (Metric) Gauge
14-819-167 Tubing Assembly; 1/4-28 TP (SR) 762 mm 18 ga
14-819-164 Tubing Assembly; 1/4-28 NT (SR) 762 mm 18 ga
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-819-167 Supplier Hamilton™ Supplier No. 88938
Add to Cart
Add to Cart
Pre-made flanged tubing assemblies made with PTFE tubing and Kel-F threaded fittings. The flanged tubing assemblies provide the perfect seal with 1/4-28 flat bottom valve ports.
Specifications
Material PTFE
Overall Length (Metric) 762 mm
Includes 0.25 to 28 flat bottom valve ports
Gauge 18 ga
Product Type Tubing Assembly; 1/4-28 TP (SR)