Learn More
Sigma Aldrich Fine Chemicals Biosciences Hamilton PRP X300 HPLC Col
Catalog No. 502908910
Encompass
The Hamilton PRP-X300 an ion exchange PSDVB/Sulfonic Acid column used in the separation of alcohols and organic acids.
Catalog No. 50-290-8910 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 82015U
The Hamilton PRP-X300 an ion exchange PSDVB/Sulfonic Acid column used in the separation of alcohols and organic acids.