Learn More
MSC Hamilton Rigid Top Plate Caster, Cast Iron, 6 in Wheel, 1400 lb Capacity
Catalog No. 5000530777
Encompass
This rigid top plate caster features a 6 inch wheel diameter and a 2 inch wheel width, crafted from durable cast iron. It offers a robust 1400 lb capacity and an overall height of 7.5 inches.
  • Plate size of 4 × 4.5 inches
  • Roller bearings
  • Hot forged construction
  • Enamel finish
  • Suitable for manufacturing applications
  • Made in the United States
Catalog No. 50-005-30777 Supplier MSC Supplier No. 75012922
This rigid top plate caster features a 6 inch wheel diameter and a 2 inch wheel width, crafted from durable cast iron. It offers a robust 1400 lb capacity and an overall height of 7.5 inches.
  • Plate size of 4 × 4.5 inches
  • Roller bearings
  • Hot forged construction
  • Enamel finish
  • Suitable for manufacturing applications
  • Made in the United States