Learn More
MSC Hamilton Swivel Top Plate Caster, 10 in Polyurethane Wheel, 3,000 lb
Catalog No. 5000531618
Encompass
This swivel top plate caster is constructed from polyurethane for high abrasion, medium moisture, and medium chemical resistance. It features a 10 in wheel diameter, a 3 in wheel width, and a robust 3,000 lb load capacity, making it suitable for demanding manufacturing applications.
  • 10 in wheel diameter and 3 in wheel width
  • Polyurethane construction offers high abrasion, medium moisture, chemical, and impact resistance with medium cushioning
  • 3,000 lb load capacity
  • 12-1/2 in overall height
  • Plate mount type with 5-1/4 x 7-1/4 in plate size
  • Precision sealed ball bearing type for the wheel
  • Hot forged kingpin swivel construction
  • Enamel finish
  • Suitable for manufacturing applications
  • Manufactured in the United States
Catalog No. 50-005-31618 Supplier MSC Supplier No. 75012997
This swivel top plate caster is constructed from polyurethane for high abrasion, medium moisture, and medium chemical resistance. It features a 10 in wheel diameter, a 3 in wheel width, and a robust 3,000 lb load capacity, making it suitable for demanding manufacturing applications.
  • 10 in wheel diameter and 3 in wheel width
  • Polyurethane construction offers high abrasion, medium moisture, chemical, and impact resistance with medium cushioning
  • 3,000 lb load capacity
  • 12-1/2 in overall height
  • Plate mount type with 5-1/4 x 7-1/4 in plate size
  • Precision sealed ball bearing type for the wheel
  • Hot forged kingpin swivel construction
  • Enamel finish
  • Suitable for manufacturing applications
  • Manufactured in the United States