Learn More
MSC Hamilton Swivel Top Plate Caster, Phenolic, 8 in Wheel Diameter, 1400 lb Capacity
Catalog No. 5000513001
Encompass
This swivel top plate caster features an 8-inch phenolic wheel with a 2-inch width, offering a 1,400 lb capacity and a 9-1/2 inch overall height. The caster is equipped with a 5 inch by 4 inch top plate, making it suitable for demanding manufacturing applications where high relative capacity and medium impact/abrasion resistance are required.
  • Wheel color is black
  • Features a roller bearing type
  • Construction is hot forged
  • Kingpin swivel construction
  • Has an enamel finish/coating
  • Suitable for manufacturing applications
  • Made in the United States
Catalog No. 50-005-13001 Supplier MSC Supplier No. 75650473
This swivel top plate caster features an 8-inch phenolic wheel with a 2-inch width, offering a 1,400 lb capacity and a 9-1/2 inch overall height. The caster is equipped with a 5 inch by 4 inch top plate, making it suitable for demanding manufacturing applications where high relative capacity and medium impact/abrasion resistance are required.
  • Wheel color is black
  • Features a roller bearing type
  • Construction is hot forged
  • Kingpin swivel construction
  • Has an enamel finish/coating
  • Suitable for manufacturing applications
  • Made in the United States