Learn More
Poltex Hamilton Tube Rack Stabilizer
SDP
Catalog No. 22000605
Change view
Click to view available options
Size:
5 Slots
Catalog No. Size
22-000-605 5 Slots
1 options
Catalog No. 22-000-605 Supplier Poltex Supplier No. SMPSTABHAM5
Add to Cart
Add to Cart
The Hamilton Tube Rack Stabilizer holds and stabilizes sample tube racks for Qiasymphony, Hamilton or other related extraction/processing machines.
  • Comes in 1, 2, 3, 4, 5, or 6 rack capacity.
  • Super durable and cleanable (alcohol resistant) PETG construction.
Specifications
For Use With (Application) Qiasymphony extractor racks, hamilton racks
Size 5 Slots