Learn More
Med Vet International 7.5-in Taylor Hammer, Chrome
Supplier:  Med Vet International 111275
Encompass_Preferred
A reflex hammer is a medical instrument used by practitioners to test deep tendon reflexes. Testing for reflexes is an important part of the neurological physical examination in order to detect abnormalities in the central or peripheral nervous system. Reflex hammers can also be used for chest percussion.
Catalog No. 50-222-3957