Learn More
Aviva Systems Biology Corp HAMP Antibody - N-terminal region (ARP41620_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41620_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HAMP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HAMP. Alternate names: HEPC, HFE2B, LEAP-1, LEAP1, PLTRApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-014-57924