Learn More
ILEX LIFE SCIENCES LLC HAMS F10 P04-12500 500 ML
SDP
Catalog No. 502733848
Encompass_Preferred
PAN-Biotech Ham's F10 Medium, w: L-Glutamine, w: 1.2 g/L NaHCO3 (500 ml). BICARBONATE: With 1.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Ham's F10 nutrient mix was developed for the serum-free culture of Chinese Hamster Ovary (CHO) cells. It can be used with many different cell types, including primary cells of rat and chicken, but also for human diploid cells. Supplementation with FBS or serum-free additives may be necessary, depending on cell type.
Catalog No. 50-273-3848 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0412500
Add to Cart
Add to Cart
PAN-Biotech Ham's F10 Medium, w: L-Glutamine, w: 1.2 g/L NaHCO3 (500 ml). BICARBONATE: With 1.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Ham's F10 nutrient mix was developed for the serum-free culture of Chinese Hamster Ovary (CHO) cells. It can be used with many different cell types, including primary cells of rat and chicken, but also for human diploid cells. Supplementation with FBS or serum-free additives may be necessary, depending on cell type.