Learn More
FUJIFILM Biosciences Hams F12/DME High Glucose 11
Catalog No. 5000450194
Encompass
Ham s F12/ DME is a 11 mixture of Ham s F12 and DMEM This extremely rich complex medium is commonly selected as the basal medium for the cultivation and study of challenging cell lines or primitive cells such as neural progenitors
Catalog No. 50-004-50194 Supplier FUJIFILM Biosciences Supplier No. 9052500ML
Ham s F12/ DME is a 11 mixture of Ham s F12 and DMEM This extremely rich complex medium is commonly selected as the basal medium for the cultivation and study of challenging cell lines or primitive cells such as neural progenitors