Learn More
FUJIFILM Biosciences Hams F12/DME High Glucose 11
Catalog No. 5000450221
Encompass
Ham s F12/ DME is a 11 mixture of Ham s F12 and DMEM This extremely rich complex medium is commonly selected as the basal medium for the cultivation and study of challenging cell lines or primitive cells such as neural progenitors
Catalog No. 50-004-50221 Supplier FUJIFILM Biosciences Supplier No. 90521L
Ham s F12/ DME is a 11 mixture of Ham s F12 and DMEM This extremely rich complex medium is commonly selected as the basal medium for the cultivation and study of challenging cell lines or primitive cells such as neural progenitors