Learn More
ILEX LIFE SCIENCES LLC HAMS F12 P04-15500 6 X 500 ML
SDP
Catalog No. 502733839
Encompass_Preferred
PAN-Biotech Ham's F12 Medium, w: stable Glutamine, w: 1.176 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 1.176 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With Stable Glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Ham's F12 nutrient mix was developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells, and mouse L-cells. It can be used for serum-free growth of CHO cultures supplemented with hormones, selenium and other supplements as well as for other mammalian cells, especially cell lines of rodent origin (particularly rabbit and rat).
Catalog No. 50-273-3839 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0415500X6
PAN-Biotech Ham's F12 Medium, w: stable Glutamine, w: 1.176 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 1.176 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With Stable Glutamine. PHENOL RED: With Phenol Red. HEPES: Without HEPES. SODIUM PYRUVATE: With sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: Ham's F12 nutrient mix was developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells, and mouse L-cells. It can be used for serum-free growth of CHO cultures supplemented with hormones, selenium and other supplements as well as for other mammalian cells, especially cell lines of rodent origin (particularly rabbit and rat).