Learn More
FUJIFILM Biosciences Hams F12K Kaighns Nutrient Mix
Catalog No. 5000450181
Encompass
Hams F12K Kaighns Nutrient Mixture F12 was originally developed for use in cloning Chinese hamster ovary andLung cells This medium is useful with serum supplements for a variety of cell culture applications including the growth of myeloma and hybridoma cells
Catalog No. 50-004-50181 Supplier FUJIFILM Biosciences Supplier No. 9077500ML
Hams F12K Kaighns Nutrient Mixture F12 was originally developed for use in cloning Chinese hamster ovary andLung cells This medium is useful with serum supplements for a variety of cell culture applications including the growth of myeloma and hybridoma cells