Learn More
Innovative Research Inc Hamster Armenian IgG Affinity Purified 50mg
Supplier:  Innovative Research Inc IHMARIGG50MG
Encompass_Preferred
Hamster Armenian IgG Affinity Purified from Innovative Research has been purified from normal Armenian Hamster Serum using Protein G affinity chromatography. This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4, with a concentration of 10.0 mg/ml and a purity of >95%.
Catalog No. 50-203-6488