Learn More
American Research Products Inc Hamster C reactiVe protein CR
Catalog No. 5000327932
Encompass
Hamster C-reactiVe protein (CRP) ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27932 Supplier American Research Products Inc Supplier No. CSBEL005991HA
Hamster C-reactiVe protein (CRP) ELISA Kit from CUSABIO Detection Range 12 5 ng/mL-800 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma tissue homogenates Method Sandwich