Learn More
Immunology Consultants Hamster (CHO) Beta 2-Microglobulin - Recombinant, 0.025mg
SDP
Catalog No. 5000435520
Encompass_Preferred
This recombinant CHO beta‑2‑microglobulin (B2M; GenBank NP_001233603.1) is expressed in CHO‑S cells with a C‑terminal 6xHIS tag and purified by nickel affinity chromatography to >90% purity. The protein appears at ~14 kDa under reducing conditions and is quantified by Bradford assay. B2M is a well‑known CHO host cell protein that frequently co‑elutes with monoclonal antibodies due to its immunoglobulin‑like domains, making it a critical impurity to monitor in biologics manufacturing. This high‑quality recombinant antigen provides consistent purity, stability, and biological relevance for ELISA development, antibody validation, and HCP assay calibration. Ideal for teams building targeted impurity detection tools or studying CHO‑derived contaminants, this B2M standard supports reliable process monitoring and enhances confidence in downstream purification strategies.
Catalog No. 50-004-35520 Supplier Immunology Consultants Supplier No. AG65004101Z
This recombinant CHO beta‑2‑microglobulin (B2M; GenBank NP_001233603.1) is expressed in CHO‑S cells with a C‑terminal 6xHIS tag and purified by nickel affinity chromatography to >90% purity. The protein appears at ~14 kDa under reducing conditions and is quantified by Bradford assay. B2M is a well‑known CHO host cell protein that frequently co‑elutes with monoclonal antibodies due to its immunoglobulin‑like domains, making it a critical impurity to monitor in biologics manufacturing. This high‑quality recombinant antigen provides consistent purity, stability, and biological relevance for ELISA development, antibody validation, and HCP assay calibration. Ideal for teams building targeted impurity detection tools or studying CHO‑derived contaminants, this B2M standard supports reliable process monitoring and enhances confidence in downstream purification strategies.