Learn More
Immunology Consultants Hamster (CHO) C-X-C Motif Chemokine 3 (CXCL3) - Recombinant, 0.025mg, Lyophilized
SDP
Catalog No. 5000435371
Encompass_Preferred
This recombinant CHO C‑X‑C Motif Chemokine 3 (CXCL3; UniProt A4URF0) is expressed in CHO cells with a C‑terminal 6xHIS tag and purified to >95% purity. CXCL3 appears at ~12 kDa under reducing conditions and is quantified by Bradford assay. As a secreted chemokine involved in neutrophil recruitment, inflammation, and cell migration, CXCL3 is a relevant CHO‑derived impurity that may impact biologics quality if not effectively removed. This high‑quality recombinant antigen provides a consistent standard for ELISA calibration, antibody development, and HCP profiling. Its CHO origin ensures biological relevance for impurity studies and process monitoring. Ideal for teams studying chemokine biology, refining purification strategies, or developing targeted HCP detection tools.
Catalog No. 50-004-35371 Supplier Immunology Consultants Supplier No. AG650447LY
This recombinant CHO C‑X‑C Motif Chemokine 3 (CXCL3; UniProt A4URF0) is expressed in CHO cells with a C‑terminal 6xHIS tag and purified to >95% purity. CXCL3 appears at ~12 kDa under reducing conditions and is quantified by Bradford assay. As a secreted chemokine involved in neutrophil recruitment, inflammation, and cell migration, CXCL3 is a relevant CHO‑derived impurity that may impact biologics quality if not effectively removed. This high‑quality recombinant antigen provides a consistent standard for ELISA calibration, antibody development, and HCP profiling. Its CHO origin ensures biological relevance for impurity studies and process monitoring. Ideal for teams studying chemokine biology, refining purification strategies, or developing targeted HCP detection tools.