Learn More
American Research Products Inc Hamster Interleukin 10 IL10
Catalog No. 5000327346
Encompass
Hamster Interleukin-10 (IL10) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27346 Supplier American Research Products Inc Supplier No. CSBEL011580HA
Hamster Interleukin-10 (IL10) ELISA kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma tissue homogenates Method Sandwich