Learn More
American Research Products Inc Hamster Interleukin 1b IL 1b
Catalog No. 5000328487
Encompass
Hamster Interleukin 1b IL-1b ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28487 Supplier American Research Products Inc Supplier No. CSBE14259HA
Hamster Interleukin 1b IL-1b ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich