Learn More
Cusabio Technology LLC HAMSTER interleukin 4 ELISA KT
Catalog No. 5000051559
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Hamster Interleukin-4 (IL4) in serum plasma and tissue homogenates using a sandwich assay format With a detection range spanning from 9 37 pg/mL to 600 pg/mL and a sensitivity of 2 34 pg/mL it provides precise measurement of IL4 concentrations The assay requires a sample volume of 50-100 L and involves an incubation period of 1-5 hours Results are determined through absorbance readings at 450 nm ensuring reliable quantification of IL4 levels in various biological samples for research applications
Catalog No. 50-000-51559 Supplier Cusabio Technology LLC Supplier No. CSBEL011659HA
This ELISA kit is designed for the quantitative detection of Hamster Interleukin-4 (IL4) in serum plasma and tissue homogenates using a sandwich assay format With a detection range spanning from 9 37 pg/mL to 600 pg/mL and a sensitivity of 2 34 pg/mL it provides precise measurement of IL4 concentrations The assay requires a sample volume of 50-100 L and involves an incubation period of 1-5 hours Results are determined through absorbance readings at 450 nm ensuring reliable quantification of IL4 levels in various biological samples for research applications