Learn More
Sigma Aldrich Fine Chemicals Biosciences Hanahans Broth SOB Medium500G
Catalog No. 502900170
Encompass
Hannahans Broth (SOB) is a microbial growth medium used for the preparation and transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides amino acids and water-soluble vitamins in a low-salt formulation. The 0.4% dextrose composition provides the E. coli carbon and energy needed to mend perforations and for replication.
Catalog No. 50-290-0170 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H8032500G
Hannahans Broth (SOB) is a microbial growth medium used for the preparation and transformation of competent cells (E. coli). This nutrient-rich microbial broth contains peptides amino acids and water-soluble vitamins in a low-salt formulation. The 0.4% dextrose composition provides the E. coli carbon and energy needed to mend perforations and for replication.