Learn More
Mopec Hand Bagging Kit
SDP
Catalog No. 22446266 Shop All Mopec Products
Change view
Click to view available options
Includes:
50 Paper Bags, 50 Rubber Bands
Catalog No. Includes
22-446-266 50 Paper Bags, 50 Rubber Bands
1 options
Catalog No. 22-446-266 Supplier Mopec Supplier No. FS477
Add to Cart
Add to Cart
Used to cover a decedent's hands in suspected suicide cases, these materials help preserve gunshot residue during transport. Paper bags and rubber bands are sealed before use to help prevent contamination, supporting protection of potential residue evidence until further examination.
Construction
  • (50) White paper bag
  • (50) Wide rubber bands
  • Packaged in a pre-sealed bag
Performance
  • Prevents loss of GSR from hands while the body is transported
Specifications
Includes 50 Paper Bags, 50 Rubber Bands