Learn More
Educational Innovations HAND BOILER
SDP
Catalog No. NC9830671
Encompass
When the handboiler is held in your hand, liquid quickly travels from the bottom bulb to the top along with numerous bubbles, giving the appearance of boiling.
Catalog No. NC9830671 Supplier Educational Innovations Supplier No. HB100
When the handboiler is held in your hand, liquid quickly travels from the bottom bulb to the top along with numerous bubbles, giving the appearance of boiling.