Learn More
Grainger Hand Crank Hose Reel 100 ft 1 1/2 ID
Catalog No. 502765704
Encompass_Preferred
The Grainger Multipurpose Hand Crank Hose Reel is a manual hose reel with a powder-coated steel frame and a zinc-plated steel reel. It has a capacity of 150 ft of 3/8 in. ID hose and includes a gooseneck that provides 180° swivel. The reel's handle folds down when not in use and it has a bracket to lock the hose at desired length. The overall dimensions are 24 in. W x 23 in. D x 41 in. H. This durable hose reel is designed for indoor or outdoor use.
Catalog No. 50-276-5704 Supplier Grainger Supplier No. 16X573
The Grainger Multipurpose Hand Crank Hose Reel is a manual hose reel with a powder-coated steel frame and a zinc-plated steel reel. It has a capacity of 150 ft of 3/8 in. ID hose and includes a gooseneck that provides 180° swivel. The reel's handle folds down when not in use and it has a bracket to lock the hose at desired length. The overall dimensions are 24 in. W x 23 in. D x 41 in. H. This durable hose reel is designed for indoor or outdoor use.