Learn More
Grainger Hand Crank Hose Reel 100 ft 3/8 ID Blck
Catalog No. 502766967
Encompass_Preferred
The COXREELS Hand Crank Hose Reel is a 100 ft capacity steel hose reel with a powder coated finish to prevent corrosion. It features a slip ring design to prevent hose kinking, a ratcheting gear drive for easy hose winding, and an adjustable hose guide. This hose reel can hold 5/8" or 3/4" ID hose up to 100 ft in length. It has a maximum working pressure of 300 PSI and is designed for air, water, lubricants, and more. The reel mounts to the wall or ceiling.
Catalog No. 50-276-6967 Supplier Grainger Supplier No. 16X560
The COXREELS Hand Crank Hose Reel is a 100 ft capacity steel hose reel with a powder coated finish to prevent corrosion. It features a slip ring design to prevent hose kinking, a ratcheting gear drive for easy hose winding, and an adjustable hose guide. This hose reel can hold 5/8" or 3/4" ID hose up to 100 ft in length. It has a maximum working pressure of 300 PSI and is designed for air, water, lubricants, and more. The reel mounts to the wall or ceiling.