Learn More
Grainger Hand Crank Hose Reel 125 ft 1/2 ID Blck
Catalog No. 502766968
Encompass_Preferred
The Coxreels Hand Crank Hose Reel is a 125 ft capacity hose reel made of steel with a powder coat finish. It features a slip ring bracket to prevent hose wear and has a gear ratio of 3.4:1 for easy hose retraction. This hose reel can hold 5/8" or 3/4" ID hose up to 125 ft in length. The reel is designed for pressures up to 300 PSI and temperatures up to 200°F. It has a folding crank handle and mounts to the wall or floor. Overall dimensions are 19.5"W x 19.5"H x 10.5"D. This heavy duty hose reel provides an easy way to neatly store longer hoses.
Catalog No. 50-276-6968 Supplier Grainger Supplier No. 16X561
The Coxreels Hand Crank Hose Reel is a 125 ft capacity hose reel made of steel with a powder coat finish. It features a slip ring bracket to prevent hose wear and has a gear ratio of 3.4:1 for easy hose retraction. This hose reel can hold 5/8" or 3/4" ID hose up to 125 ft in length. The reel is designed for pressures up to 300 PSI and temperatures up to 200°F. It has a folding crank handle and mounts to the wall or floor. Overall dimensions are 19.5"W x 19.5"H x 10.5"D. This heavy duty hose reel provides an easy way to neatly store longer hoses.