Learn More
Grainger Hand Crank Hose Reel 150 ft 1 ID Blue
Catalog No. 502767506
Encompass_Preferred
The COXREELS Hand Crank Hose Reel is a 100 ft capacity steel hose reel with a powder coated finish to prevent corrosion. It features a slip ring design to prevent hose kinking, a ratchet lock to hold hose at desired length, and an adjustable hose guide. This hose reel can hold 5/8" or 3/4" ID hose up to 100 ft in length. It has a hand crank rewind and swivel inlet fitting for ease of use. The reel dimensions are 22" diameter x 14" width with a mounting pattern of 12" x 12". It has a max pressure rating of 300 psi and max temperature rating of 150°F. This heavy duty hose reel is made in the USA.
Catalog No. 50-276-7506 Supplier Grainger Supplier No. 16X570
The COXREELS Hand Crank Hose Reel is a 100 ft capacity steel hose reel with a powder coated finish to prevent corrosion. It features a slip ring design to prevent hose kinking, a ratchet lock to hold hose at desired length, and an adjustable hose guide. This hose reel can hold 5/8" or 3/4" ID hose up to 100 ft in length. It has a hand crank rewind and swivel inlet fitting for ease of use. The reel dimensions are 22" diameter x 14" width with a mounting pattern of 12" x 12". It has a max pressure rating of 300 psi and max temperature rating of 150°F. This heavy duty hose reel is made in the USA.