Learn More
Grainger Hand Crank Hose Reel 150 ft 3/8 ID Blue
Catalog No. 502767503
Encompass_Preferred
The Grainger Multipurpose Hand Crank Hose Reel is a manual hose reel with a powder-coated steel frame and a zinc-plated steel reel. It has a capacity of 150 ft of 3/8 in. ID hose and includes a gooseneck that provides 180° swivel. The reel features a foldable crank handle for easy storage when not in use. This versatile hose reel can be mounted on the wall or used as a portable cart with two 8 in. wheels. It's designed for watering gardens, washing cars, and other general purpose applications.
Catalog No. 50-276-7503 Supplier Grainger Supplier No. 16X556
The Grainger Multipurpose Hand Crank Hose Reel is a manual hose reel with a powder-coated steel frame and a zinc-plated steel reel. It has a capacity of 150 ft of 3/8 in. ID hose and includes a gooseneck that provides 180° swivel. The reel features a foldable crank handle for easy storage when not in use. This versatile hose reel can be mounted on the wall or used as a portable cart with two 8 in. wheels. It's designed for watering gardens, washing cars, and other general purpose applications.