Learn More
Grainger Hand Crank Hose Reel 150 ft 3/8 ID Blue
Catalog No. 502766946
Encompass_Preferred
The COXREELS Hand Crank Hose Reel is a heavy duty, powder coated steel hose reel that holds up to 150 feet of 5/8 inch hose. It has a hand crank rewind and a guide arm to keep the hose neatly in place. The reel can be mounted on a wall or the included portable cart. It has a working pressure of 300 PSI and is designed for commercial and industrial applications. The reel comes with an inlet swivel and stop valve.
Catalog No. 50-276-6946 Supplier Grainger Supplier No. 16X566
The COXREELS Hand Crank Hose Reel is a heavy duty, powder coated steel hose reel that holds up to 150 feet of 5/8 inch hose. It has a hand crank rewind and a guide arm to keep the hose neatly in place. The reel can be mounted on a wall or the included portable cart. It has a working pressure of 300 PSI and is designed for commercial and industrial applications. The reel comes with an inlet swivel and stop valve.