Learn More
Grainger Hand Crank Hose Reel 200 ft 1 ID Blue
Catalog No. 502766312
Encompass_Preferred
The Coxreels Hand Crank Hose Reel is a 200 ft capacity hose reel made of steel with a powder coat finish. It features a slip ring design to prevent hose kinking, an adjustable hose guide, and a hand crank rewind with a 4:1 gear ratio for easy hose retrieval. This industrial-grade hose reel can hold 5/8" or 3/4" ID hose up to 200 ft in length. It has a max working pressure of 300 PSI and is designed for air, water, lubricants, and more. The reel comes equipped with an inlet swivel for easy hose connections.
Catalog No. 50-276-6312 Supplier Grainger Supplier No. 16X571
The Coxreels Hand Crank Hose Reel is a 200 ft capacity hose reel made of steel with a powder coat finish. It features a slip ring design to prevent hose kinking, an adjustable hose guide, and a hand crank rewind with a 4:1 gear ratio for easy hose retrieval. This industrial-grade hose reel can hold 5/8" or 3/4" ID hose up to 200 ft in length. It has a max working pressure of 300 PSI and is designed for air, water, lubricants, and more. The reel comes equipped with an inlet swivel for easy hose connections.