Learn More
Grainger Hand Crank Hose Reel 225 ft 1/2 ID Blue
Catalog No. 502767147
Encompass_Preferred
The Grainger Hand Crank Hose Reel is a 225 foot, 1/2 inch ID blue hose reel made of steel with a powder coat finish. It has a manual hand crank rewind and includes an inlet swivel and mounting hardware. This industrial grade hose reel can hold up to 225 feet of 1/2 inch ID hose and has a max pressure rating of 300 PSI. It features four non-marking rubber wheels for easy mobility. The overall dimensions are 37 inches high x 23 inches wide x 23 inches deep. This durable hose reel provides an organized storage solution for hoses up to 225 feet long.
Catalog No. 50-276-7147 Supplier Grainger Supplier No. 16X559
The Grainger Hand Crank Hose Reel is a 225 foot, 1/2 inch ID blue hose reel made of steel with a powder coat finish. It has a manual hand crank rewind and includes an inlet swivel and mounting hardware. This industrial grade hose reel can hold up to 225 feet of 1/2 inch ID hose and has a max pressure rating of 300 PSI. It features four non-marking rubber wheels for easy mobility. The overall dimensions are 37 inches high x 23 inches wide x 23 inches deep. This durable hose reel provides an organized storage solution for hoses up to 225 feet long.