Learn More
Grainger Hand Crank Hose Reel 225 ft 1/2 ID Blue
Catalog No. 502767142
Encompass_Preferred
The Coxreels Hand Crank Hose Reel is a 225 ft capacity hose reel made of steel with a powder coat finish. It features a slip ring hose guide and holds 5/8" ID hose. The reel has a hand crank rewind with a 90° elbow hose connection and swivel inlet. It can be mounted on the wall or floor and has a maximum pressure rating of 300 PSI. The reel dimensions are 26"W x 23"H x 19"D and it weighs 44 lbs.
Catalog No. 50-276-7142 Supplier Grainger Supplier No. 16X567
The Coxreels Hand Crank Hose Reel is a 225 ft capacity hose reel made of steel with a powder coat finish. It features a slip ring hose guide and holds 5/8" ID hose. The reel has a hand crank rewind with a 90° elbow hose connection and swivel inlet. It can be mounted on the wall or floor and has a maximum pressure rating of 300 PSI. The reel dimensions are 26"W x 23"H x 19"D and it weighs 44 lbs.