Learn More
Grainger Hand Crank Hose Reel 50 ft 1 1/2 ID
Catalog No. 502767145
Encompass_Preferred
The REELCRAFT Hand Crank Hose Reel is a durable steel hose reel that holds up to 105 feet of 3/4 inch ID hose. It has a powder coat finish to prevent rust and corrosion. The reel features a slow-retracting spring-loaded rewind mechanism with adjustable drag and a guide arm to keep the hose neatly in place. The hand crank allows easy extension and retraction of the hose. This heavy-duty hose reel provides an organized solution for storing lengthy hoses in commercial, industrial and residential settings.
Catalog No. 50-276-7145 Supplier Grainger Supplier No. 16X572
The REELCRAFT Hand Crank Hose Reel is a durable steel hose reel that holds up to 105 feet of 3/4 inch ID hose. It has a powder coat finish to prevent rust and corrosion. The reel features a slow-retracting spring-loaded rewind mechanism with adjustable drag and a guide arm to keep the hose neatly in place. The hand crank allows easy extension and retraction of the hose. This heavy-duty hose reel provides an organized solution for storing lengthy hoses in commercial, industrial and residential settings.