Learn More
Grainger Hand Crank Hose Reel 50 ft 3/8 ID Blue
Catalog No. 502766311
Encompass_Preferred
The COXREELS Hand Crank Hose Reel is a 50 foot capacity steel reel with powder coated finish to prevent corrosion. It features a slip ring design to prevent kinking, a ratcheting gear drive for smooth operation, and an adjustable guide arm with 8 position ratchet lock. This hose reel can hold 5/8" or 3/4" ID hose up to 300 PSI working pressure. It has a heavy duty 5" rubber non-marking wheels and is designed for commercial and industrial applications. The reel mounts to the wall or floor and is made in the USA.
Catalog No. 50-276-6311 Supplier Grainger Supplier No. 16X554
The COXREELS Hand Crank Hose Reel is a 50 foot capacity steel reel with powder coated finish to prevent corrosion. It features a slip ring design to prevent kinking, a ratcheting gear drive for smooth operation, and an adjustable guide arm with 8 position ratchet lock. This hose reel can hold 5/8" or 3/4" ID hose up to 300 PSI working pressure. It has a heavy duty 5" rubber non-marking wheels and is designed for commercial and industrial applications. The reel mounts to the wall or floor and is made in the USA.