Learn More
Thermo Scientific™ Hand Dispenser Kit
GSA_VA
Catalog No. 10451221 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Hand Dispenser Kit
Catalog No. Description
10-451-221 Hand Dispenser Kit
1 options
Catalog No. 10-451-221 Supplier Thermo Scientific™ Supplier No. 50138221
  • $2,220.00
    $1,265.65 / Each of 1
    Save $954.35
    43% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase lab flexibility with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems.
Increase flexibility in the lab with the Thermo Scientific™ Hand Dispenser Kit, compatible with many Thermo Scientific™ Barnstead™ water purification systems. This kit includes a dispenser with 3 meter cord that connects to the water system tank and a 0.2 micron final filter. It is compatible with Barnstead LabTower™, Pacific™ and Smart2Pure™ systems.
Optional for the following Barnstead water purification systems:
  • LabTower™ EDI
  • LabTower™ RO
  • LabTower™ TII
  • Smart2Pure™
  • Pacific™ TII + tank (tank must have optional pump)
  • Pacific™ RO + tank (tank must have optional pump)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Hand Dispenser Kit
For Use With (Equipment) LabTower EDI, LabTower TII, Smart2Pure, Pacific TII + Tank (Tank Must Have Optional Pump), Pacific RO + Tank (Tank Must Have Optional Pump), LabTower RO
Includes Hand Dispenser Kit (includes hand dispenser with 3 meter cord, and 0.2 micron final filter)