Learn More
Grainger Hand Drum Pump Rotary 10 gpm@120 strokes
Catalog No. 502752328
Encompass_Preferred
The Lincoln 14U105 is a hand operated rotary drum pump designed for transferring light oils and diesel fuels from drums up to 55 gallons. It features a cast iron body with Buna-N seals, a 3/4" NPT inlet and outlet, a 6" pickup tube, and a maximum flow rate of 9 GPM. This drum pump requires no electricity or batteries to operate, making it a versatile and portable fluid transfer solution. With its rugged construction, the 14U105 is built for frequent use in industrial environments when moving common petroleum-based fluids between storage drums.
Catalog No. 50-275-2328 Supplier Grainger Supplier No. 14U105
The Lincoln 14U105 is a hand operated rotary drum pump designed for transferring light oils and diesel fuels from drums up to 55 gallons. It features a cast iron body with Buna-N seals, a 3/4" NPT inlet and outlet, a 6" pickup tube, and a maximum flow rate of 9 GPM. This drum pump requires no electricity or batteries to operate, making it a versatile and portable fluid transfer solution. With its rugged construction, the 14U105 is built for frequent use in industrial environments when moving common petroleum-based fluids between storage drums.