Learn More
Grainger Hand Drum Pump Rotary 10 gpm@120 strokes
Catalog No. 502751857
Encompass_Preferred
The Lincoln 14U104 is a hand operated rotary drum pump designed for transferring light oils and diesel fuels from drums up to 55 gallons. It features a cast iron body and aluminum pump head for durability, with a 4 GPM flow rate and 25 ft maximum head. This drum pump has a 1" NPT inlet and 1" NPT outlet, an 8ft suction tube, and a 4ft discharge hose. It requires no electricity or batteries to operate. The 14U104 rotary drum pump from Lincoln provides a manual pumping solution for transferring fuels and oils from drums.
Catalog No. 50-275-1857 Supplier Grainger Supplier No. 14U104
The Lincoln 14U104 is a hand operated rotary drum pump designed for transferring light oils and diesel fuels from drums up to 55 gallons. It features a cast iron body and aluminum pump head for durability, with a 4 GPM flow rate and 25 ft maximum head. This drum pump has a 1" NPT inlet and 1" NPT outlet, an 8ft suction tube, and a 4ft discharge hose. It requires no electricity or batteries to operate. The 14U104 rotary drum pump from Lincoln provides a manual pumping solution for transferring fuels and oils from drums.