Learn More
Contec Hand Dry Wipe Dispenser Screw
Catalog No. 17010260 Shop All Contec Inc. Products
Change view
Click to view available options
Mounting:
Wall or Surface Mount
Catalog No. Mounting
17-010-260 Wall or Surface Mount
1 options
Catalog No. 17-010-260 Supplier Contec Supplier No. 9451DW
  • $258.00
    $247.65 / Each of 1
    Save $10.35
    4% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The Hand Dry Wipe Dispenser with Screw Mount offers a secure, space-saving solution for easy wipe access.
  • Downward dispenser is engineered to minimize contamination (e.g., avoid splashing from sink and dripping from hands)
  • Screw mounted to fit your needs
  • Other dispensing alternatives can be open to the environment and do not protect wipes from contamination
Specifications
Product Type Wipe Dispenser
Color Silver
Length (Metric) 29.2 cm
Width (Metric) 16.5 cm
Height (Metric) 27.9 cm
Holds Wipes
Material Stainless Steel
Mounting Wall or Surface Mount
No. of Compartments 1
Product Style Front-loading with Opening