Learn More
Grainger Crescent Nicholson Machinists File, Round, Smooth Cut, Steel, 1 ea
Catalog No. 502794850
Encompass_Preferred
This 6 in. round American pattern file features a single-cut design with teeth extending around the entire circumference. Engineered for precision material removal, this tool is primarily used for enlarging circular openings or shaping concave surfaces. Its tapered profile allows for enhanced access in slots and narrow areas. Crafted from durable steel, the file provides reliable performance across a range of materials including iron, mild steel, plastics, and wood.
  • Single-cut construction provides light material removal and produces smoother edges.
  • Circular design features teeth around the entire profile for versatile shaping.
  • Tapered shape toward the point facilitates access in confined spaces.
  • Rigid steel construction ensures durability during use.
  • Safe point helps reduce the risk of injury during operation.
  • Compatible with optional handles for improved grip and control.
Catalog No. 50-279-4850 Supplier Grainger Supplier No. 1AKA3
This 6 in. round American pattern file features a single-cut design with teeth extending around the entire circumference. Engineered for precision material removal, this tool is primarily used for enlarging circular openings or shaping concave surfaces. Its tapered profile allows for enhanced access in slots and narrow areas. Crafted from durable steel, the file provides reliable performance across a range of materials including iron, mild steel, plastics, and wood.
  • Single-cut construction provides light material removal and produces smoother edges.
  • Circular design features teeth around the entire profile for versatile shaping.
  • Tapered shape toward the point facilitates access in confined spaces.
  • Rigid steel construction ensures durability during use.
  • Safe point helps reduce the risk of injury during operation.
  • Compatible with optional handles for improved grip and control.