Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DueNorth Hand & Foot Ball Massager
Description
- Massage relief for tired achy feet
- Massage sore, tight, and tired muscles
- Rubz Ball is great for working sore hands and feet
- Rubz Ball and Rubz Massage Roller help to relieve plantar fasciitis
- Full body massage tool relieves tight and sore muscles in back, arms, shoulders, thighs and calves
Specifications
Specifications
| Color | Green |
| For Use With (Application) | Foot Massage |
| Size | Universal |
| Disposable | Reusable |
| Gender | Unisex |
| Latex Free | Yes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?