Learn More
Myron L Company HAND HELD CONDUCTIVITY METER
SDP
Catalog No. NC9513207
Encompass
4 & 5 RANGE CONDUCTIVITY METERS,SIZE: 3.4 inx4.5 inx4.0 in WEIGHT:(0.45 kg),ACCURACY: +/- 2% of full scale,REPEATABILITY: +/- 1%,TEMP COMPENSATION:50-160F (10-71 degreeC),Operate on the principle o. Please note, this item is sold with a non-dated COA.
Catalog No. NC9513207 Supplier Myron L Company Supplier No. EP10
4 & 5 RANGE CONDUCTIVITY METERS,SIZE: 3.4 inx4.5 inx4.0 in WEIGHT:(0.45 kg),ACCURACY: +/- 2% of full scale,REPEATABILITY: +/- 1%,TEMP COMPENSATION:50-160F (10-71 degreeC),Operate on the principle o. Please note, this item is sold with a non-dated COA.